Learn More
anti-ATP5F1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232325
Catalog No. NBP232325
Add to cart
Description
Description
ATP5F1 Polyclonal antibody specifically detects ATP5F1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
ATP5F1
0.5 mg/mL
Western Blot 1:1000
P24539
ATP synthase B chain, mitochondrial, ATP synthase subunit b, mitochondrial, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit b, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit b, isoform 1, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit B1, ATP synthase, H+ transporting, mitochondrial Fo complex, subunit B1, ATPase subunit b, cell proliferation-inducing protein 47, H+-ATP synthase subunit b, MGC24431, PIG47
Rabbit
IgG
Protein A purified
RUO
Primary
515
Western Blot
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
ATP5F1
This ATP5F1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 161-195 amino acids from the Central region of human ATP5F1.
28.909 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat