Learn More
ATP6AP1, Mouse, Clone: 3A2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ATP6AP1.
Supplier: Abnova Corporation H00000537M01
Description
This gene encodes a component of a multisubunit enzyme (1mDa MW) that mediates acidification of eukaryotic intracellular organelles. Vacuolar ATPase (V-ATPase) is comprised of a cytosolic V1 (site of the ATP catalytic site) and a transmembrane V0 domain. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, and receptor-mediated endocytosis. The encoded protein of this gene is approximately 45kDa and may assist in the V-ATPase-mediated acidification of neuroendocrine secretory granules. [provided by RefSeq
Sequence: SSDRDLWAPAADTHEGHITSDLQLSTYLDPALELGPRNVLLFLQDKLSIEDFTAYGGVFGNKQDSAFSNLENALDLAPSSLVLPAVDWYAVSTLTTYLQESpecifications
ATP6AP1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001183 | |
ATP6AP1 | |
ATP6AP1 (NP_001174, 51 a.a. ∼ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
3A2 | |
Mouse monoclonal antibody raised against a partial recombinant ATP6AP1. | |
ATP6AP1 | |
16A/ATP6IP1/ATP6S1/Ac45/CF2/MGC129781/VATPS1/XAP-3/XAP3 | |
Mouse | |
Affinity chromatography | |
RUO | |
537 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.