Learn More
ATPase, H+ transporting, lysosomal accessory protein 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human ATP6AP1 protein.
Supplier: Abnova Corporation H00000537D01P
Description
This gene encodes a component of a multisubunit enzyme (1mDa MW) that mediates acidification of eukaryotic intracellular organelles. Vacuolar ATPase (V-ATPase) is comprised of a cytosolic V1 (site of the ATP catalytic site) and a transmembrane V0 domain. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, and receptor-mediated endocytosis. The encoded protein of this gene is approximately 45kDa and may assist in the V-ATPase-mediated acidification of neuroendocrine secretory granules. [provided by RefSeq
Sequence: MMAAMATARVRMGPRCAQALWRMPWLPVFLSLAAAAAAAAAEQQVPLVLWSSDRDLWAPAADTHEGHITSDLQLSTYLDPALELGPRNVLLFLQDKLSIEDFTAYGGVFGNKQDSAFSNLENALDLAPSSLVLPAVDWYAVSTLTTYLQEKLGASPLHVDLATLRELKLNASLPALLLIRLPYTASSGLMAPREVLTGNDEVIGQVLSTLKSEDVPYTAALTAVRPSRVARDVAVVAGGLGRQLLQKQPVSPVIHPPVSYNDTAPRILFWAQNFSVAYKDQWEDLTPLTFGVQELNLTGSFWNDSFARLSLTYERLFGTTVTFKFILANRLYPVSARHWFTMERLEVHSNGSVAYFNASQVTGPSIYSFHCEYVSSLSKKGSLLVARTQPSPWQMMLQDFQIQAFNVMGEQFSYASDCASFFSPGIWMGLLTSLFMLFIFTYGLHMILSLKTMDRFDDHKGPTISLTQIVSpecifications
ATPase, H+ transporting, lysosomal accessory protein 1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human ATP6AP1 protein. | |
ATP6AP1 | |
16A/ATP6IP1/ATP6S1/Ac45/CF2/MGC129781/VATPS1/XAP-3/XAP3 | |
Rabbit | |
Affinity Purified | |
RUO | |
537 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001183.4 | |
ATP6AP1 | |
ATP6AP1 (NP_001174.2, 1 a.a. ∼ 470 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.