Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-ATP6V1A, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP133021
Description
ATP6V1A Polyclonal antibody specifically detects ATP6V1A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
ATP6V1A | |
0.57mg/mL | |
Western Blot 1:1000-1:20000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300 | |
523 | |
Rabbit | |
IgG | |
Polyclonal | |
Immunogen affinity purified | |
RUO | |
Human,Mouse |
Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunoprecipitation,Immunohistochemistry (Paraffin) | |
Unconjugated | |
Affinity Purified | |
70kD, isoform 1, ATP6A1, ATP6V1A1ATPase, H+ transporting, lysosomal, subunit A1, ATPase, H+ transporting, lysosomal (vacuolar proton pump), alpha polypeptide, ATPase, H+ transporting, lysosomal 70kDa, V1 subunit A, EC 3.6.3, EC 3.6.3.14, H(+)-transporting two-sector ATPase, subunit A, H+-transporting ATPase chain A, vacuolar (VA68 type), HO68, VA68, vacuolar ATP synthase catalytic subunit A, ubiquitous isoform, Vacuolar ATPase isoform VA68, vacuolar proton pump alpha subunit 1, Vacuolar proton pump subunit alpha, V-ATPase 69 kDa subunit, V-ATPase 69 kDa subunit 1, V-ATPase A subunit 1, V-ATPase subunit A, Vma1, VPP2, V-type proton ATPase catalytic subunit A | |
ATP6V1A | |
Recombinant protein encompassing a sequence within the center region of human ATP6V1A. The exact sequence is proprietary. | |
68 kDa | |
Primary | |
0.1 ml | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
Tested in human. Not yet tested in any other species. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only