Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6V1E1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP215519
Description
ATP6V1E1 Polyclonal specifically detects ATP6V1E1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ATP6V1E1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
ATP6E2ATP6V1E, ATP6E31kDa subunit, ATPase, H+ transporting, lysosomal (vacuolar proton pump) 31kD, ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E isoform 1, ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E1, H+-transporting ATP synthase chain E, vacuolar, p31, Vacuolar proton pump subunit E 1, V-ATPase 31 kDa subunit, V-ATPase subunit E 1, V-ATPase, subunit E, Vma4, V-type proton ATPase subunit E 1 | |
Rabbit | |
26 kDa | |
0.1 mL | |
Primary | |
Human, Mouse. Expected to cross react based on sequence identity: Chicken (85%), Rat (98%), Bovine (97%), Xenopus tropicalis (83%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.17mg/mL | |
Western Blot 1:500-1:3000 | |
P36543 | |
ATP6V1E1 | |
Recombinant protein encompassing a sequence within the center region of human ATP6V1E1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
529 | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction