Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V-ATPase E2 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA529206

Catalog No. PIPA529206


Add to cart

Description

Description

PA5-29206 targets ATP6V1E2 in WB applications and shows reactivity with mouse samples. The PA5-29206 immunogen is recombit fragment corresponding to a region within amino acids 1 and 226 of Human ATP6V1E2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Subunit of the peripheral V1 complex of vacuolar ATPase essential for assembly or catalytic function. V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells. This isoform is essential for energy coupling involved in acidification of acrosome.
Specifications

Specifications

V-ATPase E2
Polyclonal
Unconjugated
Atp6v1e2
Liquid
ATP6V1E2
Recombit fragment corresponding to a region within amino acids 1 and 226 of Human ATP6V1E2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Western Blot
0.69 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q9D593
ATP6E1, ATP6EL2, ATP6V1EL2, VMA4, V-ATPase subunit E 2, V-type proton ATPase subunit E 2, vacuolar proton pump subunit E 2, vacuolar-type proton-translocating ATPase subunit E1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
74915
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit