Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ATP6V1H, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232203

Catalog No. NBP232203


Add to cart

Description

Description

ATP6V1H Polyclonal antibody specifically detects ATP6V1H in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

ATP6V1H
1.0 mg/mL
Western Blot 1:500 - 1:2500, Immunoprecipitation 2 - 10 ug/mg lysate
NP_057025.2
ATPase, H+ transporting, lysosomal 50/57kD, V1 subunit H, ATPase, H+ transporting, lysosomal 50/57kDa, V1 subunit H, CGI-11, MSTP042, NBP1, Nef-binding protein 1, Protein VMA13 homolog, SFD, SFDalpha, SFDbeta, vacuolar ATP synthase subunit H, vacuolar ATPase subunit H, vacuolar proton pump H subunit, Vacuolar proton pump subunit H, Vacuolar proton pump subunit SFD, V-ATPase 50/57 kDa subunits, V-ATPase H subunit, V-ATPase subunit H, VMA13, V-type proton ATPase subunit H
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
ATP6V1H
The immunogen this antibody was made to, maps to a region between residue 400 to 450 of human ATPase, H+ Transporting, Lysosomal 50/57kDa, V1 Subunit H using the numbering given in entry NP_057025.2 (GeneID 51606).
Immunogen affinity purified
RUO
Primary
51606
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only