Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V-ATPase H Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525106

Catalog No. PIPA525106

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and porcine based on sequence homology.

ATP6V1H encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c ft., c ft., and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene encodes the regulatory H subunit of the V1 domain which is required for catalysis of ATP but not the assembly of V-ATPase.
Specifications

Specifications

V-ATPase H
Polyclonal
PBS with 0.09% sodium azide
Q9UI12, Q8BVE3
MSTP042, NBP1, SFD, SFDalpha, SFDbeta, VMA13, V-ATPase 50/57 kDa subunits, V-ATPase H subunit, V-ATPase subunit H, V-type proton ATPase subunit H, nef-binding protein 1, protein VMA13 homolog, vacuolar ATP synthase subunit H, vacuolar ATPase subunit H, vacuolar proton pump H subunit, vacuolar proton pump subu
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
108664, 51606
Western Blot
Unconjugated
ATP6V1H
Liquid
ATP6V1H
KLH conjugated synthetic peptide between 400-426 amino acids from the C-terminal region of human ATP6V1H
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit