Learn More
ATP7B Rabbit anti-Human, Rat, DyLight® 550, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522771
Catalog No. PIPA522771
Add to cart
Description
Description
This antibody does not react with mouse samples.
ATP7b is an important protein for copper transport and elimination of excess copper from the body. ATP7b transports metals in and out of cells using ATP. There are 3 known isoforms of the ATP7b gene; A is found in the liver, kidney, and brain, the shorter form B is found in brain tissue, and the third isoform, known as WND/140 KDA is found in mitochondria. Mutations in the ATP7b gene can cause Wilson ft.s disease, an inherited disorder causing copper poisoning in the brain and liver.
Specifications
Specifications
ATP7b
Polyclonal
DyLight™ 550
ATP7B
Liquid
ATP7B
A synthetic peptide made to an internal sequence nearer the C-terminus of human ATP7b.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
sodium borate with no preservative
P35670, Q64535
RP11-327P2.1, PWD, WC1, WD, WND, ATPase, Cu(2+)- transporting, beta polypeptide, Wilson disease-associated protein, copper pump 2, copper-transporting ATPase 2
Rabbit
IgG
100 μL
4° C, store in dark
Primary
24218, 540