Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP7B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513429

Catalog No. PIPA513429


Add to cart

Description

Description

ATP7B is a member of the P-type cation transport ATPase family and a protein with several membrane-spanning domains, an ATPase consensus sequence, a hinge domain, a phosphorylation site, and at least 2 putative copper-binding sites. This protein functions as a monomer, exporting copper out of the cells, such as the efflux of hepatic copper into the bile.
Specifications

Specifications

ATP7B
Polyclonal
PBS with 0.09% sodium azide
P35670, Q64446
Copper-transporting ATPase 2; Copper pump 2; Wilson disease-associated protein; WND/140 kDa; WND; WC1; PWD; ATP7B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11979, 540
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ATP7B
Liquid
ATP7B
KLH conjugated synthetic peptide between 1361-1391 amino acids from the C-terminal region of human ATP7B
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit