Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ATPAF1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP215524

Catalog No. NBP215524


Add to cart

Description

Description

ATPAF1 Polyclonal specifically detects ATPAF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ATPAF1
0.89mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Q5TC12
ATP synthase mitochondrial F1 complex assembly factor 1, ATP11 homolog, ATP11ATP11p, Atp11p, FLJ22351, homolog of yeast ATP11, MGC88060
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
64756
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Affinity Purified
ATPAF1
Recombinant protein encompassing a sequence within the center region of human ATPAF1. The exact sequence is proprietary.
36 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human. Expected to cross react based on sequence identity: Mouse (92%).
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only