Learn More
ATP13A2 Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA118321
Catalog No. 01-670-574
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-18321 detects ATPase from rat samples. PA1-18321 has been successfully used in immunohistochemistry procedures. The PA1-18321 immunogen is a synthetic peptide corresponding to residues C-E(147) L H R Q E E A K Q V L R Y Y(161) of mouse ATP13A2, conjugated to KLH. Reconstitute with 100 μL of distilled water.
Specifications
Specifications
ATP13A2
Polyclonal
whole serum with no preservative
Lyophilized
ATP13A2
Synthetic peptide corresponding to residues C-E(147) L H R Q E E A K Q V L R Y Y(161) of mouse ATP13A2.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
362645
Immunohistochemistry
Unconjugated
Atp13a2
Probable cation-transporting ATPase 13A2, EC 3.6.3., ATP13A2
Rabbit
IgG
RUO
Antibody
Polyclonal
Rat