Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AU1 Tag Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA126548
Description
PA1-26548 detects AU1 tag. PA1-26548 has been successfully used in immunocytochemistry, ELISA, and Western blot procedures. The PA1-26548 immunogen is purified, full length, native AU1: DTYRYI conjugated to KLH.
AU1 is a commonly used epitope tag engineered onto the N- or C- terminus of a protein of interest so that the tagged protein can be analyzed and visualized by immunochemical methods. The recognized AU1 sequence represents the amino acid sequence DTYRYI, derived from the major capsid protein of bovine papillomavirus-1 (BPV-1).Specifications
AU1 tag | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4° C | |
Liquid | |
Tag |
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
AU1 Epitope Tag | |
Purified, full length, native AU1: DTYRYI conjugated to KLH. | |
100 μg | |
Primary | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction