Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AU5 Tag, HRP, Polyclonal, Invitrogen™

Catalog No. PIPA126596
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA126596 100 μg
1 options

Catalog No. PIPA126596

Supplier: Thermo Scientific PA126596

Goat Polyclonal Antibody

AU5 is a commonly used epitope tag engineered onto the N- or C- terminus of a protein of interest so that the tagged protein can be analyzed and visualized by immunochemical methods. The recognized AU5 sequence represents the amino acid sequence TDFYLK.

PA1-26596 detects AU5 tag.PA1-26596 has been successfully used in and Western blot procedures.The PA1-26596 immunogen is a synthetic peptide, conjugated to KLH, corresponding to residues T D F Y L K of AU5 tag.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AU5 tag
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate HRP
Gene Alias AU5 Epitope Tag
Host Species Goat
Immunogen Synthetic peptide: TDFYLK conjugated to KLH
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C
Product Type Antibody
Form Purified
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.