Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AU5 Tag Rabbit anti-Tag, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA126552

Catalog No. PA126552


Add to cart

Description

Description

PA1-26552 detects AU5 tag. PA1-26552 has been successfully used in immunocytochemistry and Western blot procedures. The PA1-26552 immunogen is purified, full length, native AU5: TDFYLK conjugated to KLH.

AU5 is a commonly used epitope tag engineered onto the N- or C- terminus of a protein of interest so that the tagged protein can be analyzed and visualized by immunochemical methods. The recognized AU5 sequence represents the amino acid sequence TDFYLK.
Specifications

Specifications

AU5 tag
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
100 μg
4° C
Primary
Tag
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
AU5 Epitope Tag
Purified, full length, native AU5: TDFYLK conjugated to KLH.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit