Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AUTS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP132784
Description
AUTS2 Polyclonal antibody specifically detects AUTS2 in Human, Mouse, Zebrafish samples. It is validated for Western Blot.Specifications
AUTS2 | |
Polyclonal | |
Unconjugated | |
PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
autism susceptibility candidate 2, autism susceptibility gene 2 protein, autism-related protein 1, KIAA0442FBRSL2, MGC13140 | |
Rabbit | |
139 kDa | |
100 μL | |
Primary | |
Human and Mouse. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1.0mg/mL | |
Western Blot 1:500-1:3000 | |
Q8WXX7 | |
AUTS2 | |
Recombinant protein encompassing a sequence within the center region of human AUTS2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
26053 | |
Human, Mouse, Zebrafish | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction