Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-AUTS2, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128462

Catalog No. NBP128462


Add to cart

Description

Description

AUTS2 Polyclonal antibody specifically detects AUTS2 in Human, Mouse, Canine samples. It is validated for Peptide ELISA.
Specifications

Specifications

AUTS2
0.5 mg/mL
Peptide ELISA Detection limit 1:32000
NP_056385.1
autism susceptibility candidate 2, autism susceptibility gene 2 protein, autism-related protein 1, KIAA0442FBRSL2, MGC13140
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize isoforms 1 and 2 (NP_056385.1 and NP_001120703.1 resp.).
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
AUTS2
Peptide with sequence C-ERPPSHTLKDIEAR corresponding to C-Terminus according to NP_056385.1, NP_001120703.1.
Immunogen affinity purified
RUO
Primary
26053
Canine, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only