Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avian Influenza HA Type 2 Goat anti-Virus, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA520293

Catalog No. PIPA520293


Add to cart

Description

Description

A suggested positive control is Hemagglutinin HA1 recombit protein. PA5-20293 can be used with blocking peptide PEP-0410.

Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus was identified in specimens obtained from patients in Mexico and the United States. The virus spread quickly around the world and on June 11, 2009, the World Health Organization declared it a pandemic. Influenza A virus has one of sixteen possible Hemagglutinin (HA) surface proteins and one of nine possible Neuraminidase (NA) surface proteins. The Hemagglutinin protein facilitates viral attachment while Neuraminidase is involved in viral release. These proteins also elicit immune responses that prevent infection or independently reduce viral replication. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin. The distinct antigenic properties of the new swine influenza virus compared with seasonal influenza A (H1N1) virus suggest that human immunity against new swine influenza virus is limited, although the age distribution of reported cases suggests some degree of protection in older age groups.
Specifications

Specifications

Avian Influenza HA Type 2
Polyclonal
Unconjugated
Liquid
Goat
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
Virus
ELISA, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Influenza A Hemagglutinin; Avian Hemagglutinin 2; Influenza HA
A synthetic peptide corresponding to 12 amino acids near the amino terminus of the Hemagglutinin protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit