Learn More
Avidin, Biotin, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA126846
Catalog No. PA126846
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The biological functin of avidin is not known. It is a homotetrameric glycoprotein found in chicken egg white, and forms a strong non covalent specific complex with biotin (one molecule of biotin per subunit of avidin).
PA1-26846 detects Avidin. PA1-26846 will not react with streptavidin.PA1-26846 has been successfully used in immunohistochemistry (paraffin) (frozen), ELISA and Western blot procedures.The PA1-26846 immunogen is Avidin from hen egg white.
Specifications
Specifications
Avidin
Polyclonal
Biotin
AVD
Purified
AVD
Avidin from hen egg white.
IgG fraction
RUO
Antibody
Polyclonal
Many
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot
10mg/mL
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% sodium azide; pH 7.2
P02701
AVD, avID
Rabbit
IgG
1mg
Store at -20°C
Primary
396260