Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avidin Mouse anti-Chicken, Clone: A3G7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA121417

Catalog No. MA121417


Add to cart

Description

Description

MA1-21417 detects Avidin in Chicken samples. MA1-21417 is specific for egg white avidin. No cross reaction is seen with human and animal serum proteins and streptavidin. The antibody reacts with avidin without interfering with its biotin-binding activity. MA1-21417 has been successfully used in ELISA and Western Blot procedures. The predicted molecular weight of MA1-21417 is 17 kDa. MA1-21417 immunogen corresponds to Egg white avidin. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The biological function of avidin is not known. It is a homotetrameric glycoprotein found in chicken egg white, and forms a strong non-covalent specific complex with biotin (one molecule of biotin per subunit of avidin).
Specifications

Specifications

Avidin
Monoclonal
7 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P02701
AVD, avID
Mouse
IgG1
500 μg
4° C
Primary
396260
ELISA, Immunohistochemistry, Western Blot
A3G7
Unconjugated
AVD
Liquid
AVD
Egg white avidin.
Ion-Exchange Chromatography
RUO
Antibody
Monoclonal
Chicken
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit