Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AVPI1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524008

Catalog No. PIPA524008


Add to cart

Description

Description

This antibody is predicted to react with bovine based on sequence homology.

May be involved in MAP kinase activation, epithelial sodium channel (ENaC) down-regulation and cell cycling.
Specifications

Specifications

AVPI1
Polyclonal
PBS with 0.09% sodium azide
Q5T686
PP5395, RP11-548K23.7, VIP32, VIT32, AVP-induced protein 1, arginine vasopressin-induced protein 1, vasopressin-induced transcript
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
60370
Flow Cytometry, Western Blot
Unconjugated
AVPI1
Liquid
AVPI1
KLH conjugated synthetic peptide between 56-83 amino acids from the N-terminal region of human AVPI1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit