Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Axin-1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131013

Catalog No. NBP131013


Add to cart

Description

Description

Axin-1 Polyclonal antibody specifically detects Axin-1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Axin-1
0.76mg/mL
Western Blot 1:500-1:3000
Affinity Purified
AXIN1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Axin 1. The exact sequence is proprietary.
96 kDa
0.1 ml
Apoptosis, Wnt Signaling Pathway
Primary
8312
Human
Western Blot
Unconjugated
PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal
axin 1, axin-1, AXINaxis inhibitor 1, Axis inhibition protein 1, fused, mouse, homolog of, hAxin, MGC52315
Rabbit
IgG
Antigen Affinity-purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Xenopus laevis (85%). Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only