Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Axl APC-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
Supplier: R&D Systems FAB8541A
This item is not returnable.
View return policy
Description
Axl Monoclonal specifically detects Axl in Mouse samples. It is validated for Flow Cytometry.Specifications
Axl | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
558 | |
Mouse | |
Purified |
Flow Cytometry | |
175128 | |
Flow Cytometry 10 uL/10^6 cells | |
Q00993 | |
AXL | |
Mouse myeloma cell line NS0-derived recombinant mouse Axl Ala19-Trp445 Accession # Q00993 | |
100 Tests | |
Primary | |
Detects mouse Axl. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction