Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Axl, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP122964

Catalog No. NBP122964

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

Axl
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation (Negative)
NP_068713.2
AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL
Rabbit
IgG
100 ul
Cell Cycle and Replication
Primary
558
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
AXL
The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human AXL receptor tyrosine kinase using the numbering given in entry NP_068713.2 (GeneID 558).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only