Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axotrophin, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA128120

Catalog No. PIPA128120

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

PA1-28120 detects Axotrophin from human samples.PA1-28120 has been successfully used in Western blot applications.The PA1-28120 immunogen is: Recombinant fragment, corresponding to amino acids 481-680 of Human Axotrophin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Axotrophin
Polyclonal
MARCH7
AXO, AXOT, DKFZp586F1122, MARCH-VII, RNF177
Chicken
Antigen affinity chromatography
RUO
64844
Store at -20°C, Avoid Freeze/Thaw Cycles
Purified
Western Blot
Unconjugated
Q9H992
MARCH7
Recombinant fragment, corresponding to amino acids 481-680 of Human Axotrophin.
50 μg
Primary
Human
Antibody
IgY
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit