Learn More
antizyme inhibitor 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human AZIN1 protein.
Supplier: Abnova Corporation H00051582D01P
Description
Ornithine decarboxylase (ODC) catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. Ornithine decarboxylase antizymes play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. The protein encoded by this gene is highly similar to ODC. It binds to ODC antizyme and stabilizes ODC, thus inhibiting antizyme-mediated ODC degradation. Two alternatively spliced transcript variants have been found for this gene. [provided by RefSeq
Sequence: MKGFIDDANYSVGLLDEGTNLGNVIDNYVYEHTLTGKNAFFVGDLGKIVKKHSQWQNVVAQIKPFYTVKCNSAPAVLEILAALGTGFACSSKNEMALVQELGVPPENIIYISPCKQVSQIKYAAKVGVNILTCDNEIELKKIARNHPNAKVLLHIATEDNIGGEEGNMKFGTTLKNCRHLLECAKELDVQIIGVKFHVSSACKESQVYVHALSDARCVFDMAGEIGFTMNMLDIGGGFTGTEFQLEEVNHVISPLLDIYFPEGSGVKIISEPGSYYVSSAFTLAVNIIAKKVVENDKFPSGVEKTGSDEPAFMYYMNDGVYGSFASKLSEDLNTIPEVHKKYKEDEPLFTSSLWGPSCDELDQIVESCLLPELNVGDWLIFDNMGADSFHEPSAFNDFQRPAIYYMMSFSDWYEMQDAGITSDSMMKNFFFVPSCIQLSQEDSFSAEASpecifications
antizyme inhibitor 1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human AZIN1 protein. | |
AZIN1 | |
MGC3832/MGC691/OAZI/OAZIN/ODC1L | |
Rabbit | |
Affinity Purified | |
RUO | |
51582 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_015878.4 | |
AZIN1 | |
AZIN1 (NP_056962.2, 1 a.a. ∼ 448 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.