Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B Cells Mouse anti-Rat, Clone: 69-IB3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA140234

Catalog No. 01-676-571


Add to cart

Description

Description

MA1-40234 detects B-cell specific antigen from rat samples.MA1-40234 has been successfully used in flow cytometry and immunohistochemistry (frozen) applications.The MA1-40234 immunogen is WAG/Rij cells.
Specifications

Specifications

B Cells
Monoclonal
0.1 mg/mL
PBS with 50% glycerol and 0.01% sodium azide
Mouse
IgG1
100 μg
4° C
Primary
Rat
Flow Cytometry, Immunohistochemistry (Frozen)
69-IB3
Unconjugated
Liquid
WAG/Rij cells.
Purified
RUO
Antibody
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit