Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B-Raf Mouse, Unlabeled, Clone: 13, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612375

 View more versions of this product

Catalog No. BDB612375


Add to cart

Description

Description

The Raf kinase family, A-Raf, B-Raf, and C-Raf, act as downstream effectors for Ras. These ser/thr kinases have homologous kinase domains referred to as conserved regions (CR1, CR2, and CR3). Rafs have a critical role in the response to many growth factors including: EGF, PDGF, insulin, IL-2, IL-3, CSF-1, and GM-CSF. Raf can directly interact with Ras-GTP and subsequently become activated. This leads to Raf activation of the MAP kinase pathway by phosphorylating and activating MEK, which then phosphorylates and activates MAP kinaes (ERK).
A-Raf is a proto-onocogene that is highly expressed in urogenital and kidney tissues while B-Raf is expressed at high levels in cerebrum and testes. C-Raf appears to be uniformly and ubiquitously expressed. Cell transformation by A-Raf has been shown to be independent of p21ras, suggesting the possibility of an independent signal transduction pathway. B-Raf is expressed as multiple isoforms that range from 69-72kDa and 79-99kDa. In hematopoietic cells, B-Raf is activated by EPO, GM-CSF, and SCF. Thus, the Raf kinases are important Ras effectors involved in growth factor signaling in a wide variety of cells and tissues.

Western Blotting

Specifications

Specifications

B-Raf
13
Unconjugated
Affinity Purified
Human B-Raf aa. 285-406
150μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.