Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-B220/CD45R, Alexa Fluor 594, Clone: RA3-6B2, R&D Systems™
Rat Monoclonal Antibody
Specifications
Antigen | B220/CD45R |
---|---|
Clone | RA3-6B2 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1217T025
|
R&D Systems
FAB1217T025 |
25 Tests | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
FAB1217T100
|
R&D Systems
FAB1217T100 |
100 Tests | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Specifications
B220/CD45R | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Supernatant | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
5788 | |
Abelson mouse leukemia virus-induced pre-B tumor cells | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
RA3-6B2 | |
Flow Cytometry | |
Alexa Fluor 594 | |
Rat | |
Mouse | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
PTPRC | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects mouse B220/CD45R. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title