Learn More
anti-B220/CD45R, Alexa Fluor 647, Clone: RA3-6B2, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB1217R025
 View more versions of this product
Catalog No. FAB1217R025
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
B220/CD45R
RA3-6B2
Flow Cytometry 5 uL/10^6 cells
Supernatant
PTPRC
Abelson mouse leukemia virus-induced pre-B tumor cells
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5788
Mouse
Flow Cytometry
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Rat
IgG2a
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects mouse B220/CD45R.