Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

B3GNT4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524189

Catalog No. PIPA524189

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a member of the beta-1,3-N-acetylglucosaminyltransferase protein family. The encoded enzyme is involved in the biosynthesis of poly-N-acetyllactosamine chains and prefers lacto-N-neotetraose as a substrate. It is a type II transmembrane protein.
Specifications

Specifications

B3GNT4
Polyclonal
PBS with 0.09% sodium azide
Q9C0J1, Q1RLK6
B3GN-T4, beta3Gn-T4, BGnT-4, beta-1,3-Gn-T4, beta-1,3-N-acetylglucosaminyltransferase 4, beta-1,3-N-acetylglucosaminyltransferase bGn-T4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
231727, 79369
Flow Cytometry, Western Blot
Unconjugated
B3GNT4
Liquid
B3GNT4
KLH conjugated synthetic peptide between 233-260 amino acids from the central region of human B3GNT4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit