Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B3GNT7 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524723

Catalog No. PIPA524723

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

B3GNT7 is a glycosyltransferase which may play a role in elongation of the keratan sulfate carbohydrate backbone.
Specifications

Specifications

B3GNT7
Polyclonal
PBS with 0.09% sodium azide
Q8NFL0, Q8K0J2
beta3GnT7, BGnT-7, beta 1,3-N-acetylglucosaminyltransferase 7, beta-1,3-Gn-T7, beta-1,3-N-acetylglucosaminyltransferase 7, beta3Gn-T7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
227327, 93010
Immunohistochemistry, Western Blot
Unconjugated
B3GNT7
Liquid
B3GNT7
KLH conjugated synthetic peptide between 256-282 amino acids from the central region of human B3GNT7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit