Learn More
B4GALT6 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524190
Catalog No. PIPA524190
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This antibody is predicted to react with mouse based on sequence homology.
This gene is one of seven beta-1,4-galactosyltransferase (beta4GalT) genes. They encode type II membrane-bound glycoproteins that appear to have exclusive specificity for the donor substrate UDP-galactose; all transfer galactose in a beta1,4 linkage to similar acceptor sugars: GlcNAc, Glc, and Xyl. Each beta4GalT has a distinct function in the biosynthesis of different glycoconjugates and saccharide structures. As type II membrane proteins, they have an N-terminal hydrophobic signal sequence that directs the protein to the Golgi apparatus and which then remains uncleaved to function as a transmembrane anchor. By sequence similarity, the beta4GalTs form four groups: beta4GalT1 and beta4GalT2, beta4GalT3 and beta4GalT4, beta4GalT5 and beta4GalT6, and beta4GalT7. The enzyme encoded by this gene is a lactosylceramide synthase important for glycolipid biosynthesis.
Specifications
Specifications
B4GALT6
Polyclonal
Unconjugated
B4GALT6
Liquid
B4GALT6
KLH conjugated synthetic peptide between 319-346 amino acids from the C-terminal region of human B4GALT6
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
Q9UBX8
B4Gal-T6, beta4Gal-T6, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 6, UDP-Gal:glucosylceramide beta-1,4-galactosyltransferase, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 6, beta-1,4-GalTase 6, beta-1,4-galactosyltransferase 6, beta4GalT-VI
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9331