Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-B7-2/CD86, Alexa Fluor 647, Clone: 37301, R&D Systems™

Mouse Monoclonal Antibody

Specifications

Antigen B7-2/CD86
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human B7-2/CD86
Clone 37301
Applications Flow Cytometry
Conjugate Alexa Fluor 647
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB141R025
View Documents
R&D Systems
FAB141R025
25 Tests This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
FAB141R100
View Documents
R&D Systems
FAB141R100
100 Tests This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Specifications

Specifications

B7-2/CD86
37301
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Monoclonal
Primary
Detects human B7-2/CD86 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) B7-1, recombinant mouse B7-2, recombinant rat B7-2, rhB7-H1, rhB7-H2, rhB7-H3, rhB7-H3b, rhB7-H4, or rhB7-L2 is observed.
Protein A or G purified from ascites
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human B7-2/CD86
Flow Cytometry
Ascites
Mouse
IgG1
942
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit