Learn More
anti-B7-2/CD86, PerCP, Clone: 37301, R&D Systems™
Manufacturer:  R&D Systems FAB141C025
 View more versions of this product
Catalog No. FAB141C025
Add to cart
Specifications
Specifications
B7-2/CD86
37301
Flow Cytometry 10 uL/10^6 cells
Purified
CD86
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human B7-2/CD86
Protein A or G purified from ascites
RUO
Primary
942
Human
Flow Cytometry
PerCP
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86
Mouse
IgG1
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human B7-2/CD86 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) B7-1, recombinant mouse B7-2, recombinant rat B7-2, rhB7-H1, rhB7-H2, rhB7-H3, rhB7-H3b, rhB7-H4, or rhB7-L2 is observed.