Learn More
anti-B7-H1/PD-L1, Fluorescein, Polyclonal, R&D Systems™
Manufacturer:  R&D Systems FAB1019F100
 View more versions of this product
Catalog No. FAB1019F100
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
PD-L1/B7-H1
Fluorescein
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Affinity Purified
CD274
Mouse myeloma cell line NS0-derived recombinant mouse B7-H1/PD-L1, Phe19-Thr238, Accession # Q9EP73
Antigen Affinity-purified
RUO
Primary
29126
Mouse
Flow Cytometry
Flow Cytometry 10 uL/10^6 cells
Q9EP73
B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Goat
IgG
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Polyclonal
Detects mouse B7-H1/PD-L1 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human B7-H1/PD-L1 is observed.