Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

anti-B7-H3/CD276, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232251

Catalog No. NBP232251



Description

Description

B7-H3/CD276 Polyclonal antibody specifically detects B7-H3/CD276 in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

B7-H3/CD276
1.0 mg/mL
Immunoprecipitation 2 - 10 ug/mg lysate
NP_001019907.1
B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
CD276
The immunogen this antibody was made to, maps to a region between residue 484 to 534 of human CD276 Molecule using the numbering given in entry NP_001019907.1 (GeneID 80381).
Immunogen affinity purified
RUO
Primary
80381
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only