Learn More
MilliporeSigma™ anti-BACE1, C-Terminal (485-501), Polyclonal
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ 195111100UL
Catalog No. 19-511-1100UL
Add to cart
Description
Description
Specifically detects BACE1, C-Terminal (485-501) Clone: in Human, Mouse, Rat samples, and it is validated for Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Radioimmunoassay
β-secretase has been identified as a novel membrane-bound aspartyl protease named BACE1 (β-site amyloid precursor protein cleaving enzyme 1). It is one of two endopeptidases that cleave theβ-amyloid precursor protein (APP) resulting in the accumulation of theβ-amyloid deposits involved in the development of Alzheimer's disease.
Specifications
Specifications
BACE1, C-Terminal (485-501)
Unconjugated
Undiluted Serum
a synthetic peptide (CLRQQHDDFADDISLLK) corresponding to amino acids 485-501 of human BACE1
100μL
−20°C
Polyclonal
Human, Murine, Rat
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Radioimmunoassay
Undiluted serum.
Rabbit
IgG
RUO
Primary
Recognizes both the immature and mature forms of the BACE1 protein.