Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BACE1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA112529
Description
In Western blot, this antibody detects a ∽70 kDa band in human brain lysate.
Accumulation of the amyloid-b (Ab) plaque in the cerebral cortex is a critical event in the pathogenesis Alzheimer′s disease. Ab peptide is generated by proteolytic cleavage of the b-amyloid protein precursor(APP) at b- and g-sites by two proteases. APP is first cleaved by b-secretase, producing a soluble derivative of the protein and a membrane anchored 99-amino acid carboxy-terminal fragment (C99). The C99 fragment serves as substrate for g-secretase to generate the 4 kDa amyloid-b peptide, which is deposited in the brains of all sufferers of Alzheimer′s disease. The long-sought b-secretase was recently identified by several groups independently and designated beta-site APP cleaving enzyme (BACE) and aspartyl protease 2 (Asp2). bACE/Asp2 is a novel transmembrane aspartic protease and colocalizes with APP.Specifications
BACE1 | |
Polyclonal | |
Unconjugated | |
BACE1 | |
Liquid | |
BACE1 | |
Synthetic peptide corresponding to residues 485-501 of human BACE. | |
Antigen affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P56817, P56818 | |
beta Secretase 1 | |
Rabbit | |
IgG | |
100 μg | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
23621, 23821 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title