Learn More
BACE2 Ab-1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BACE2.
Supplier: Abnova Corporation PAB9773
Description
Cerebral deposition of amyloid beta peptide is an early and critical feature of Alzheimer's disease and a frequent complication of Down syndrome. Amyloid beta peptide is generated by proteolytic cleavage of amyloid precursor protein by 2 proteases, one of which is the protein encoded by this gene. This gene localizes to the 'Down critical region' of chromosome 21. The encoded protein, a member of the peptidase A1 protein family, is a type I integral membrane glycoprotein and aspartic protease. Three transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Specifications
BACE2 Ab-1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of BACE2. | |
In PBS (0.02% sodium azide) | |
AEPLC/ALP56/ASP1/ASP21/BAE2/CDA13/CEAP1/DRAP | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
BACE2 | |
BACE2 | |
A synthetic peptide corresponding to amino acids 44-59 of human BACE2. | |
RUO | |
25825 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.