Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BACE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112530

Catalog No. 01-674-190


Add to cart

Description

Description

In Western blot, this antibody detects a ~70 kDa band in human heart tissue lysate.

BACE2 or Beta Secretase 2 is responsible for the proteolytic processing of the amyloid precursor protein (APP). BACE2 cleaves APP between residues 690 and 691. This cleavage leads to the generation and extracellular release of beta-cleaved soluble APP. BACE2 is also know to cleave APP between residues 671 and 672. Accumulation of APP is a signature of Alzheimer disease and frequent complication of adult Down syndrome patients.
Specifications

Specifications

BACE2
Polyclonal
Unconjugated
BACE2
Liquid
BACE2
Synthetic peptide corresponding to residues 44-59 of human BACE2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9Y5Z0, Q9JL18, Q6IE75
BACE2, AEPLC, ALP56, ASP1, ASP21, BAE2, CDA13, CEAP1, DRAP, beta secretase 2
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
25825, 288227, 56175
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit