Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BACH2, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523642

Catalog No. PIPA523642

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

BACH2
Polyclonal
Unconjugated
BACH2
Liquid
BACH2
KLH conjugated synthetic peptide between 815-841 amino acids from the C-terminal region of human BACH2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.5mg/mL
PBS with 0.09% sodium azide
Q9BYV9
BTB and CNC homolog 2, transcription regulator protein BACH2
Rabbit
IgG
400μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
60468
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit