Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bad Mouse, Unlabeled, Clone: 48, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610392

 View more versions of this product

Catalog No. BDB610392


Add to cart

Description

Description

Isolated by screening for Bcl-2 interacting proteins, Bad shows significant homology to Bcl-2 within the Bcl-2 homology domains 1 and 2 (BH1 and BH2). In addition, several other proteins involved in cell death such as Bax, Bcl-X[L], Mcl-1, and A1 share similar homology with Bcl-2. Bcl-2 is known to oppose several apoptotic signals and is considered to be a central downstream cell death repressor. Bcl-X[L] represses apoptosis, but its short form, Bcl-X[S], promotes cell death. Bax is known to homodimerize as well as heterodimerize with Bcl-2. An excess concentration of Bax opposes the ability of Bcl-2 to repress cell death. Bad can selectively dimerize with Bcl-X[L] and Bcl-2, but not with Bax, Bcl-X[S], Mcl-1, A1, or itself. In mammalian cells, Bad binds more strongly to Bcl-X[L] than Bcl-2. This may explain why Bad reverses the death repressor activity of Bcl-X[L], but not that of Bcl-2. The formation of the Bad-Bcl-X[L] heterodimer displaces Bax andrestores favorable conditions for apoptosis.

This antibody is tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

Bad
48
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA
150μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.