Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bad Mouse anti-Human, Mouse, Clone: BYC001, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05605
Description
This Anti-Bad Antibody, clone BYC001 is validated for use in immunoprecipitation, western blotting for the detection of Bad.Specifications
Bad | |
Monoclonal | |
Unconjugated | |
NM_032989 | |
Mouse | |
Protein G chromatography | |
Primary | |
2-8°C for one year | |
IgG |
Immunoprecipitation, Western Blot | |
BYC001 | |
0.1M Tris-glycine, pH 7.4, 0.15M NaCl and 0.05% sodium azide. | |
BAD, BBC2, BBC6, BCL2L8 | |
Full-length murine Bad containing a N-terminal His6-tag | |
200 μg | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction