Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAF155/170 Mouse anti-Human, Clone: DXD12, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04082
Description
This Anti-BAF155/170 Antibody, clone DXD12 is validated for use in western blotting for the detection of BAF155/170.
Specifications
BAF155/170 | |
Monoclonal | |
Unconjugated | |
NM_003074.2 | |
Mouse | |
Protein A purfied | |
Primary | |
−20°C for 24 months. | |
IgG |
Western Blot | |
DXD12 | |
100μg of Protein A purified mouse monoclonal IgG in 100μL of 1X PBS, pH 7.0, 0.1% Azide. | |
SMARCC1, CRACC1, SRG3, BAF155, SWI3, Rsc8 | |
Synthetic peptide corresponding to residues 591-608 of human BAF155. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction