Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAF53A, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA129471

Catalog No. PIPA129471

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a family member of actin-related proteins (ARPs), which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene encodes a 53kDa subunit protein of the BAF (BRG1/brm-associated factor) complex in mammals, which is functionally related to SWI/SNF complex in S. cerevisiae and Drosophila; the latter is thought to facilitate transcriptional activation of specific genes by antagonizing chromatin-mediated transcriptional repression. Together with beta-actin, it is required for maximal ATPase activity of BRG1, and for the association of the BAF complex with chromatin/matrix. Three transcript variants that encode two different protein isoforms have been described.

PA1-29471 detects BAF53A from mouse, human samples.PA1-29471 has been successfully used in Western blot applications.The PA1-29471 immunogen is: Recombinant fragment, corresponding to amino acids 1-429 of Human BAF53A.
Specifications

Specifications

BAF53A
Polyclonal
Unconjugated
ACTL6A
Purified
ACTL6A
Recombinant fragment, corresponding to amino acids 1-429 of Human BAF53A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Western Blot
1mg/mL
PBS with no preservative
O96019, Q9Z2N8
ACTL6, ARPN-BETA, Arp4, BAF53A, INO80K, MGC5382
Chicken
IgY
50μg
Store at -20°C
Primary
86
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit