Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAFF/BLyS/TNFSF13B Rat anti-Mouse, Clone: 121808, R&D Systems™

Rat Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen BAFF/BLyS/TNFSF13B
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse BAFF Ala127-Leu309 Accession # Q9WU72
Clone 121808
Applications Western Blot, Flow Cytometry, CyTOF
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1357SP
View Documents
R&D Systems
MAB1357SP
25 ug
Each for $114.50
Add to cart
 
MAB1357100
View Documents
R&D Systems
MAB1357100
100 ug
Each for $377.00
Add to cart
 
MAB1357500
View Documents
R&D Systems
MAB1357500
500 ug
Each for $839.00
Add to cart
 
Description

Description

BAFF/BLyS/TNFSF13B Monoclonal antibody MAB1357 Specifically detects BAFF/BLyS/TNFSF13B in Mouse samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

BAFF/BLyS/TNFSF13B
121808
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Rat
IgG2a
10673
RUO
Mouse myeloma cell line NS0-derived recombinant mouse BAFF Ala127-Leu309 Accession # Q9WU72
Western Blot, Flow Cytometry, CyTOF
Purified
Q9WU72
Monoclonal
Primary
Detects mouse BAFF in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse (rm) 4-1BB Ligand, rmEDA, rmFas Ligand, rmOX40 Ligand, rmTNF-alpha , rmTRANCE, rmTWEAK, recombinant human (rh) APRIL, rhBAFF, rhEDA-A2, rhGITR Ligand, rhLIGHT, rhLymphotoxin alpha 1/beta 2, rhLymphotoxin alpha 2/beta 1, rhTNF-alpha , rhTRAIL, rhVEGI, recombinant canine, cotton rat, equine, feline, porcine, or rat TNF-alpha .
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit