Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAG-1 Mouse anti-Human, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611868

Catalog No. BDB611868


Add to cart

Description

Description

Members of the Bcl-2 protein family function to inhibit (Bcl-2, Bcl-XL, Mcl-1, A1) or promote (Bax, Bak, Bcl-Ls, Bad, Bid, Bim, Bik) apoptosis. Bcl-2- associated athanogene-1 (BAG-1) was identified through its interaction with Bcl-2 family proteins. BAG-1 protein has been reported to be expressed as at least three isoforms, 50 kDa (BAG-1L), 46 kDa (BAG-1M/RAP46), and 33 kDa (BAG-1S), which contain conserved C-terminal regions that have an Hsp70-binding domain (HBD). BAG-1M was also identified as RAP46, a protein that binds steroid hormone receptors. BAG1-M is transported to the nucleus along with the ligand-bound glucocorticoid receptor (GR), where it downregulates GR binding to DNA in an Hsp70-dependent manner. In the Hsp70-Hip chaperone system, BAG-1M/RAP46 acts as a negative regulator of Hsp70 chaperone activity, while in the Hsc70-dj systems, BAG-1 acts as a positive cochaperone. BAG-1 mRNA is ubquitously expressed in cells and tissues, while BAG-1L protein is preferentially expressed in many tumor cell lines. Thus, BAG-1 isoforms are multifunctional proteins that may be involved in protein folding, hormone receptor signaling, and malignant transformation.

Specifications

Specifications

BAG-1
19
Unconjugated
Affinity Purified
Human BAG-1 aa. 155-258
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit