Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAG4 Rabbit anti-Bovine, Canine, Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141498

Catalog No. 01-675-323


Add to cart

Description

Description

Suggested positive control: Hela whole cell extract, HeLa cell lysate.

Several cell surface receptors that are involved in apoptosis contain intracellular death domains and are capable of triggering apoptosis when activated by their respective ligands (1). Due to presence of these death domains and presence of various stimuli in the serum as well as overexpression leading to receptor oligomerization, which leads to constitutive activation of the apoptotic pathway, it has been extremely difficult to generate permanent cell lines overexpressing specific receptors in culture. However, in normal states, this death domain activity is silenced presumably due to presence of other cellular factors.  SODD (Silencer of Death Domain), a DR3 associated protein, was isolated by yeast two-hybrid interaction assays (2). SODD associates with the intracellular domains of TNF-R1, but not with those of TNF-R2, Fas, DR4, or DR5.  Overexpression has been found to suppress the TNF induced cell death as well as NF-kB activation (2). SODD may act as a negative regulatory protein that is normally associated with death domain of TNF-R1 and probably inhibit self-oligomerization properties of death domain and maintain TNF-R1 in an inactive, monomeric state.
Specifications

Specifications

BAG4
Polyclonal
Unconjugated
BAG4
Liquid
BAG4
Synthetic peptide corresponding to residues S(2) A L R R S G Y G P S D G P S(16) of human SODD/BAG4.
Protein G
RUO
Antibody
Polyclonal
Bovine, Canine, Human, Mouse, Non-human primate, Rat
Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
O95429, Q8CI61
BAG-4, SODD
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
361167, 516353, 607954, 67384, 9530
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit