Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BAG5, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP133300

Catalog No. NBP133300

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

BAG5
0.68mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
BAG5
Recombinant protein encompassing a sequence within the center region of human BAG5. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
BAG family molecular chaperone regulator 5, BAG-5, BAG-family molecular chaperone regulator-5, Bcl-2-associated athanogene 5, BCL2-associated athanogene 5, KIAA0873
Rabbit
IgG
0.1 ml
Apoptosis
Primary
9529
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only